VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-1170EA 1620 DKK
115-1170 115-1159 115-1171 115-1169 115-1168 115-1167 115-1166 115-1165 115-1164 115-1163 115-1160 115-1174 115-1158 115-1157 115-1156 115-1155 115-1154 115-1153 115-1152 115-1151 115-1150 115-1161 115-1172 115-1173 115-1179 115-1178 115-1177 115-1176 115-1175 115-1149 115-1217 115-1228 115-1227 115-1226 115-1225 115-1224 115-1223 115-1222 115-1221 115-1220 115-1206 115-1218 115-1231 115-1216 115-1215 115-1214 115-1212 115-1211 115-1210 115-1209 115-1208 115-1219 115-1241 115-1257 115-1256 115-1255 115-1254 115-1249 115-1248 115-1247 115-1246 115-1245 115-1244 115-1229 115-1242 115-1230 115-1240 115-1239 115-1238 115-1237 115-1236 115-1235 115-1234 115-1233 115-1232 115-1205 115-1243 115-1207 115-1182 115-1258 115-1192 115-1204 115-1203 115-1202 115-1201 115-1200 115-1199 115-1198 115-1197 115-1196 115-1195 115-1180 115-1191 115-1193 115-1181 115-1190 115-1189 115-1188 115-1187 115-1186 115-1185 115-1184 115-1183
VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth
Måtter Belastningsreducerende Måtter
These mats are made of low density polyurethane providing an ergonomic, anti-fatigue profile. Bubble mats are designed for dry, non critical environments, reducing trip hazards, body aches, quality issues and sick leave. The smooth, flat mats are ideal for dry, non critical environments where sitting and standing work areas are combined, or where carts or trolleys must be used.
  • Moulded bevelled edges will not curl up, thus eliminating trip hazards
  • Any size or any shape - will always be one piece: No overlapping or worn out connecting mats
  • Ergonomic design