VWR®, Mats, Cleanroom Anti-Fatigue, Bubble and Smooth
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-0745EA 1300 DKK
115-0745 115-0746 115-0747 115-0748 115-0749 115-0757 115-0759 115-0760 115-0756 115-0732 115-0736 115-0731 115-0733 115-0730 115-0735 115-0727 115-0737 115-0738 115-0739 115-0740 115-0741 115-0715 115-0734 115-0743 115-0728 115-0744 115-0726 115-0725 115-0724 115-0723 115-0722 115-0721 115-0720 115-0719 115-0718 115-0717 115-0716 115-0742 115-0758 115-1082 115-1093 115-1092 115-1091 115-1090 115-1089 115-1088 115-1087 115-1086 115-1085 115-1083 115-1081 115-1080 115-1079 115-1078 115-1147 115-1084 115-1070 115-1122 115-1121 115-1120 115-1077 115-1076 115-1075 115-1074 115-1073 115-1124 115-1071 115-1125 115-1069 115-1068 115-1067 115-1066 115-1065 115-1064 115-1063 115-1062 115-1061 115-1072 115-1136 115-1096 115-1146 115-1145 115-1144 115-1143 115-1142 115-1141 115-1140 115-1139 115-1123 115-1137 115-1148 115-1135 115-1134 115-1133 115-1132 115-1131 115-1130 115-1129 115-1128 115-1127 115-1126 115-1138 115-1094 115-1108 115-1119 115-1118 115-1117 115-1116 115-1115 115-1114 115-1113 115-1112 115-1111 115-1109 115-1107 115-1106 115-1105 115-1104 115-1103 115-1099 115-1098 115-1110 115-1095
VWR®, Mats, Cleanroom Anti-Fatigue, Bubble and Smooth
Måtter Belastningsreducerende Måtter
These mats offer the comfort of a polyurethane mat which is suitable for ISO Class 5 cleanroom (Class 100) environments. It is either available with a bubble surface, that features a unique ergonomic and anti-fatigue design, or with a smooth surface that is ideal for areas that combine sitting and standing.
  • Extremely good chemical and UV resistance (except for organic solvents)
  • Free from silicone and latex
  • Closed cell/non porous surface allows for easy wash-down and cleaning
  • Smooth mat: Ideal if carts or trolleys must be used
  • Bubble mat: Ergonomic