Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

11460.FIEA 885 DKK
11460.FI 11460.GH
Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Tin

Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈1.8g/50x50mm Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Kogepunkt: 2270 °C (1013 hPa)
Smeltepunkt: 232 °C
Massefylde: 7,31 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR