Cleanroom frocks, Tyvek® IsoClean®, IC270B, Bulk
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IC270BWH0B/M IC270BWH0B/S IC270BWH0B/L IC270BWH0B/XL IC270BWH0B/2X IC270BWH0B/3X
114-9438EA 2580 DKK
114-9438 114-9439 114-9437 114-9440 114-9435 114-9436
Cleanroom frocks, Tyvek® IsoClean®, IC270B, Bulk
Engangsbeklædning
These frocks deliver an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Clean-processed