Intelligent sole cleaning machine (ISCM)
Leverandør: GUANG DONG SUOREC TECHNOLOG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CLW-T10P CLW-T18P CLW-T28P CLW-T13P
113-9164EA 27680 DKK
113-9164 113-9166 113-9167 113-9165
Intelligent sole cleaning machine (ISCM)
Skosværte og børster
The Intelligent Sole Cleaning Machine (ISCM) effectively prevents embarrassing footprints by eliminating the need for sticky mat or shoe cover inventory, disposal, and replacement costs.
  • High particle removal efficiency up to 96%
  • Simple installation, and easy to clean
  • User-friendly and environmentally-friendly design
  • Long service life up to 8 years
  • Improve product yield and save cleanroom maintenance costs
Simple cleaning operation requires stepping onto the sole cleaning machine and remaining atop the cleaning grills for 3 seconds. Then step onto theMore Product Information