Bouffant cap, BioClean™ BBC
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BBC-W
113-8790EA 1210 DKK
113-8790
Bouffant cap, BioClean™ BBC
Engangsbeklædning
These lightweight cleanroom bouffant caps are made of non woven PP.
  • Hypoallergenic, soft fabric
  • Elasticated double stitching
  • Latex-free
  • Compatible with a variety of environments
Density: 10 g/m²
Bestil nu
Specifikationer
Environmentally Preferable
Køn
Outfit applikationer Cleanroom