Cleanroom Overalls Tyvek® IsoClean® model IC183B DS
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IC183BWHDS/M IC183BWHDS/L IC183BWHDS/XL IC183BWHDS/2X IC183BWHDS/3X IC183BWHDS/S IC183BWHDS/XS
113-8629EA 3970 DKK
113-8629 113-8630 113-8631 113-8632 113-8633 113-8628 DUPOIC183BWHDS/XS
Cleanroom Overalls Tyvek® IsoClean® model IC183B DS
Engangsbeklædning
These unhooded overalls are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Ultra-low linting fabric
  • Unhooded overall with bound neck
  • Tyvek® covered elasticated thumb loops
  • Tunnelled elastication at wrists and ankles; stitched-in elasticated waist
  • Front zipper closure with storm flap