VWR® Cleanroom Garment Sets
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-7875EA 2970 DKK
113-7875 113-7876 113-7877 113-7878 113-7879 113-7880
VWR® Cleanroom Garment Sets
Engangsbeklædning
This single-use garment set includes overall, hood, boot covers and sleeve protectors, all made from breathable SMS fabric.
  • Overall with collar, raglan-style sleeves, two-way zip fastener and thumb loop (latex-free)
  • Overall features include bound seams and elasticated wrists, ankles and back for tighter fit and easier donning
  • Overall is folded, with snap release to arms and legs to aid aseptic donning
  • Boot covers with anti-skid PU sole
  • Low particle shedding
  • Garments are gamma sterilised to SAL 10⁻⁶
These sterile garments are ideal for use in cleanroomMore Product Information