Laboratoriekittel, Tyvek® 500
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYPL30SWHLG53P TYPL30SWHMD53P TYPL30SWHXL53P TYPL30SWH2X53P TYPL30SWHLG52P TYPL30SWHMD52P TYPL30SWHSM005 TYPL30SWHXL52P TYPL30SWH2X52P TYPL30SWHLG5NP TYPL30SWHMD5NP TYPL30SWHXL5NP TYPL30SWH2X5NP
113-0467EA 4030 DKK
113-0467 113-0468 113-0469 113-0470 113-0471 113-0472 113-3882 113-0473 113-0474 113-0475 113-0476 113-0477 113-0478
Laboratoriekittel, Tyvek® 500
Engangsbeklædning
These garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Fabric is ultra-low linting
  • With collar and stitched internal seams
  • Anti-static treatment (EN 1149-5) on both sides
  • PL30 with straight cuffs, PL309 + PL30NP with elasticated cuffs (not tunnelled)
  • Available with zip or press stud closure, with
More Product Information
Bestil nu
Specifikationer
Model PL30, 3 pockets PL309, 2 pockets PL30NP
Environmentally Preferable
Lukkestørrelse Med trykknapper Med lynlås Med trykknapper
Lommer med lommer med lommer uden lommer
Køn Unisex Unisex Unisex
Outfit applikationer Chemical protection,Laboratory Chemical protection,Laboratory Chemical protection,Laboratory
Weight 41 g/m² 41 g/m² 41 g/m²