Kemisk resistent beskyttelsesdragt, Tyvek® 500 Industry
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYCCF5SWHSM010 TYCCF5SWHMD010 TYCCF5SWHLG010 TYCCF5SWHXL010 TYCCF5SWH2X010 TYCCF5SWH3X010
113-3853EA 2820 DKK
113-3853 113-3854 113-3855 113-3856 113-3857 113-9700
Kemisk resistent beskyttelsesdragt, Tyvek® 500 Industry
Engangsbeklædning
These Tyvek® overalls are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size).
  • Ultra-low linting fabric
  • Collar, elasticated wrists, waist and ankles
  • Tyvek® zip and zip flap for enhanced protection
  • Gusset for improved fit
  • Stitched internal seams to help reduce contamination from the inside to the outside of the