VWR® Maximum, Beskyttelsesdragter til renrum
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1076EA 1450 DKK
113-1076 113-1075 113-1074 113-1079 113-1086 113-1089 113-1077 113-1087 113-1078 113-1085 113-1084 113-1082 113-1081 113-1080 113-1088 113-9161
VWR® Maximum, Beskyttelsesdragter til renrum
Engangsbeklædning
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
  • Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
  • Inset sleeves, zip closure, elasticated cuffs
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding:
More Product Information