Chemical resistant boot covers, Tychem® 2000 C, model POBA
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYCPOBASYL00
113-0878EA 2650 DKK
113-0878
Chemical resistant boot covers, Tychem® 2000 C, model POBA
Engangsbeklædning
Specially designed for use with Tychem® apparel, these boot covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Knee-length boot covers with slip-retardant sole and fixing ties
  • Sole is partially stitched: splash-proof, not fully liquid-tight
  • Anti-static treatment (EN 1149-5) on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against aMore Product Information