White nitrile gloves, sterile, Kimtech™ G3
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
56888 56889 56890 56891 56892 56893 56894 56887
112-6391EA 3110 DKK
112-6391 112-6392 112-6393 112-6394 112-6395 112-6396 112-6397 112-6390
White nitrile gloves, sterile, Kimtech™ G3
Handsker
Kimtech™ G3 sterile white nitrile gloves provide enhanced sterile cleanroom protection for a wide range of contaminants. With a textured finish and skin-friendly nitrile composition, the hand-specific sterile gloves protect users and processes alike.
  • Sterility Assurance Level (SAL) 10⁻⁶
  • Certificate of analysis available online
  • Certificate of sterility available online
  • Fully sterilised white nitrile material is stronger and leaner than latex
  • Certified, high levels of contamination protection against particles, micro-organisms, viruses, cytotoxic drugs, and