Cleanroom gloves, polychloroprene, BioClean Fusion™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BFAP-XL BFAP-XS BFAP-S BFAP-M BFAP-L BFAP-XXL S-BFAP-1010 S-BFAP-5055 S-BFAP-6065 S-BFAP-7075 S-BFAP-8085 S-BFAP-9090
112-3836EA 5060 DKK
112-3836 112-3837 112-3835 112-3834 112-3499 112-3838 112-3894 112-3895 112-3896 112-3897 112-3898 112-3899
Cleanroom gloves, polychloroprene, BioClean Fusion™
Handsker
These ambidextrous gloves with beaded cuffs provide excellent grip. The non sterile version is suitable for chemotherapy applications (ASTM D 6978-05 tested).
  • Sterile or non sterile, powder-free, textured finish
  • Latex-free to eliminate Type 1 latex allergies
  • Beaded cuff provides easier donning and stability on the arm
  • Polychloroprene provides exceptional tactility
  • Suitable for double donning
  • Excellent ESD properties
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 374-1/2/3, EN 455; Cat. III, AQL 1,5
Certificeringer: Manufactured in aMore Product Information