Full face masks with Typhoon™ valve, Force®10
Leverandør: JSP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BPB003-004-000 BPB003-104-000 BPB003-204-000
111-2600EA 2000 DKK
111-2600 111-2601 111-2602
Full face masks with Typhoon™ valve, Force®10
Åndedrætsværn Genanvendelige masker Fuldtdækkende ansigtsmasker
The Force®10 Typhoon™ is an extremely lightweight, easy to maintain, full face mask with panoramic visor. With 3 available sizes (S,M,L) and the ability to fit PressToCheck™ filters, the user can feel reassured of an excellent fit.
  • Available in Small, Medium, or Large sizes for a perfect fit
  • Compatible with PressToCheck™ Filters (not included)
  • Typhoon™ exhalation valve for reduced breathing resistance
  • Polycarbonate lens, ABS buckles
  • Engineered for easy maintenance
  • One of the lightest full face masks on the market
The Force®10 Typhoon™ has beenMore Product Information