Autoclavable goggles
Leverandør: UNIVET
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611.S0.00.00 611.S0.00.01 611.S1.00.00 611.S1.00.01
111-2349EA 2310 DKK
111-2349 111-2428 111-2429 111-2430
Autoclavable goggles
Beskyttelsesbriller
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
  • Available with direct or indirect ventilation
  • Elastic band in anti-static silicone with quick and smooth system
  • RX insert
  • Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
Certificeringer: EN 166.
Leveringsinformation: More Product Information