Bump caps, AirPro™
Leverandør: CENTURION SAFETY PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S38NBRP S38RB S53SCHVY S38G S38E S38K S38KRP S38NB S38PNB S38R
111-1684EA 218 DKK
111-1684 111-1687 CENUS53SCHVY 111-1677 111-1676 111-1680 111-1682 111-1683 111-1685 111-1686
Bump caps, AirPro™
Hjelme Bump Caps
This cap features an inner ABS shell and a breathable polyester cover with PTFE coating. Suitable applications include automotive, airline, transport, shipping, railway and engineering industries.
  • Cap material is water resistant
  • Proban version features flame retardant properties
  • Side mesh panels and breathable fabric provide improved airflow and increased coolness
  • Lower nape strap depth for optimum head retention
  • Reflective features for improved safety and higher visibility
  • Available with reduced peak for improved upward visibility or close work