Reusable half masks, MX/PF
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1001558
111-0548EA 620 DKK
111-0548
Reusable half masks, MX/PF
Åndedrætsværn Genanvendelige masker Halvmasker
The half mask MX/PF is designed to work when the type of protection depends on the cartridges connected, gases, particles or combined. The APF is evaluated from 12 to 50.
  • Easy assembly
  • Silicone facepiece for additional comfort and easy decontamination
  • Triple flange seal
  • Wide, horizontal skin contact with pleated design
  • Good fit accross a wide variety of facial geometries
The half mask SPERIAN MX PF can be used only with certified T SERIES cartridges (following the standard EN 141 , EN 143)
Recommended for long term use