Engangspartikelfiltrerende åndedrætsværn, FFP3, 8833
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8833
111-0451EA 1080 DKK
111-0451
Engangspartikelfiltrerende åndedrætsværn, FFP3, 8833
Åndedrætsværn Engangsmasker Partikelmasker
The robust shell and cup design of the mask conforms well to most face shapes and sizes, and maintains its shape during use. The excellent fit is provided by the convex shape, the nose clip, and the twin-strap design.
  • Cool Flow™ valve reduces heat build-up to offer comfortable protection, particularly in hot and humid conditions
  • Soft inner face-seal ring provides secure face fit and increases wearer comfort
  • Reliable, effective protection against fine particulates
  • Comfortable, lightweight off-the-face design
  • Red colour coded straps denoting FFP3 protection