Tin ≥99.995% (metals basis), powder -100 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

11013.04 11013.09 11013.18 11013.30
11013.04EA 508 DKK
11013.04 11013.09 11013.18 11013.30
Tin ≥99.995% (metals basis), powder -100 mesh
Tin

Purity: 99.995% CAS No.: 7440-31-5 Appearance: Grey Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Kogepunkt: 2270 °C (1013 hPa)
Smeltepunkt: 232 °C
Massefylde: 7,31 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR