Magnesiumoxid 99,998 Metals basis, Puratronic®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Magnesia usta

10800.18 10800.09 10800.30
10800.18EA 6010 DKK
10800.18 10800.09 10800.30
Magnesiumoxid 99,998 Metals basis, Puratronic®
Magnesium oxide

Purity: 99.998% CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: -22 Mesh Powder Density: 3.58 Refractive Index: 1.736 Odour: Odourless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109

Formel: MgO
Molekylvægt: 40.3 g/mol
Kogepunkt: 3600 °C
Smeltepunkt: 2852 °C
Massefylde: 3.58
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011109
CAS-nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR