Water ≥99.9%, LiChrosolv®, PFAS grade, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydrogen oxide

MQ100

1047351000
1047351000.EA 843 DKK
1047351000.
Water ≥99.9%, LiChrosolv®, PFAS grade, Supelco®
Vand

Water tested for EPA533 and EPA537.1 PFAS
methods LiChrosolv®.



  • ESI/APCI (+) <2 ppb; ESI/APCI (-) <10 ppb

  • Lowest impurity profile for Interference free baseline
  • Borosilicate glass bottles minimize contamination with metal ions
  • Lowest levels of trace metal impurities: for minimised metal ion adduct formation <5ppb

  • Microfiltration through 0.2 μm filter


LiChrosolv ultra-high purity solvents for PFAS testing are specially designed for this high demanding application. The products will help to reduce background interference and will not contain any of the PFAS compounds above the limit values defined by the EPA 533 and EPA 537.1 methods. This allows reliable and effective testing to facilitate identification and quantitation of PFAS analytes and eliminates the risk of false positive or false negative results in LC-MS/MS analysis. PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.


Formel: H₂O
Molekylvægt: 18,02 g/mol
Kogepunkt: 100,0 °C at 1.013 hPa
Smeltepunkt: 0 °C
Massefylde: 1,000 g/cm³
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5

Bestil nu

Specifikationer Testresultater

EPA 533/537.1 Tested Conforms
UHPLC Gradient (at 230 nm) ≤2.0 mAU
Evaporation Residue ≤0.7 mg/l
Titratable Acid (meq/g) ≤0.001
Titratable Base (meq/g) ≤0.0002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR