Tantal ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

10350.G1 10350.G5 10350.G9
10350.G1EA 870 DKK
10350.G1 10350.G5 10350.G9
Tantal ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Tantalum

Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈13.0g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011252

Formel: Ta
Molekylvægt: 180.95 g/mol
Kogepunkt: 5427
Smeltepunkt: 2996
Massefylde: 16.69 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011252
CAS-nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR