Tantal (50 ppm Nb) ≥99.97% (metals basis), powder, Puratronic® -325 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

10345.14 10345.22 10345.36 10345.88
10345.14EA 2260 DKK
10345.14 10345.22 10345.36 10345.88
Tantal (50 ppm Nb) ≥99.97% (metals basis), powder, Puratronic® -325 mesh
Tantalum

Purity: 99.97% CAS No.: 7440-25-7 Appearance: Grey Odour: Odourless UN No.: 3089 MDL No.: MFCD00011252

Formel: Ta
Molekylvægt: 180.95 g/mol
Kogepunkt: 5427
Smeltepunkt: 2996
Massefylde: 16.6 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011252
CAS-nummer: 7440-25-7
EINECS: 231-135-5
UN nummer: 3089
ADR: 4.1 II
Merck Index: 12,09223

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR