Hæmatoxylin solution (Mayer), Q Path® for microscopy

Leverandør: VWR Q-Path Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: DcH-48, Natural Black I, Hydroxybrasilin

10047005.EA 677 DKK
10047005. 10047105.
Hæmatoxylin solution (Mayer), Q Path® for microscopy
Hæmatoxylinfarvningsopløsning

Ready to use solution for HES histological staining

Formel: C₁₆H₁₄O₆
Molekylvægt: 302.28 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00078111
CAS-nummer: 517-28-2
IVDLogo

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Colour of solution Purplish red
Density (20/4) 1,05 - 1,07
pH (20°C) 2,2 - 2,6
Absorbance (0.1 %) 559 - 561 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR