Rhodiumstandard, 1g/L Rh i salpetersyre (from Rh(NO₃)₃) Certipur® enkeltelement standard til ICP, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.70345.0100
1.70345.0100EA 2312.87 DKK
1.70345.0100
Rhodiumstandard, 1g/L Rh i salpetersyre (from Rh(NO₃)₃) Certipur® enkeltelement standard til ICP, Supelco®
Standarder Kationiske standarder Rhodium Standards
Certificeringer: Traceable to SRM from NIST