Eddikesyre 1 mol/l (1 N), Titripur®, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

MQ100

1.60305.1000
1.60305.1000EA 286 DKK
1.60305.1000
Eddikesyre 1 mol/l (1 N), Titripur®, Supelco®
Eddikesyre
Formel: H₃CCOOH
Molekylvægt: 60.05 g/mol
Kogepunkt: 118
Smeltepunkt: 17
Flammepunkt: 38,5
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR