(±)-α-Methoxyphenyleddikesyre Reag. Ph. Eur., Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

M-Clarity™ quality level = MQ200

1.59712.0010
1.59712.0010EA 858 DKK
1.59712.0010
(±)-α-Methoxyphenyleddikesyre Reag. Ph. Eur., Supelco®
(±)-α-Methoxyphenyleddikesyre
Formel: C₉H₁₀O₃
Kogepunkt: 165 °C (24 hPa)
Smeltepunkt: 69…71 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064217
CAS-nummer: 7021-09-2
EINECS: 230-300-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR