HPLC columns, Purospher® STAR RP-18 endcapped
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
1.50036.0001EA 5850 DKK
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
HPLC columns, Purospher® STAR RP-18 endcapped
Kromatografikolonner
Fast method development of complex samples across the pH range from 1,5 to 10,5 with different mobile phases and temperature conditions.
Purospher® STAR RP-18 endcapped HPLC columns are designed for universal use. Basic, neutral and metal-chelating compounds can easily be separated with simple mobile phases – naturally without peak tailing. Thanks to its outstanding performance and stability, Purospher® STAR RP-18 endcapped offers maximum flexibility in method development. Robust methods can be developed over the entire pH range from 1,5 to 10,5. High pH-stability allows the separation ofMore Product Information