HPLC columns, SeQuant® ZIC®-cHILIC
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50662.0001 1.50670.0001 1.50658.0001 1.50659.0001 1.50656.0001 1.50657.0001 1.50660.0001 1.50661.0001 1.50669.0001 1.50658.1003
1.50662.0001EA 11440 DKK
1.50662.0001 1.50670.0001 1.50658.0001 1.50659.0001 1.50656.0001 1.50657.0001 1.50660.0001 1.50661.0001 1.50669.0001 1.50658.1003
HPLC columns, SeQuant® ZIC®-cHILIC
Kromatografikolonner
ZIC®-cHILIC is based on the same technology platform as ZIC®-HILIC, but has a different functional group and an improved manufacturing procedure. The different charge orientation of the zwitterionic functional group leads to a different – and complementary – selectivity for polar hydrophilic compounds. This selectivity is especially beneficial for hydrophilic compounds that are negatively charged, positively charged, zwitterionic or neutral, for example, organic acids, amino acids, metabolites, aminoglycosides and nucleotides.
  • Complementary selectivity by reoriented charges in