HPLC columns, Purospher® STAR Si and NH₂
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50244.0001 1.50245.0001 1.50247.0001 1.50248.0001 1.50249.0001 1.50267.0001 1.50268.0001 1.50269.0001 1.50356.0001 1.50357.0001
1.50244.0001EA 5260 DKK
1.50244.0001 1.50245.0001 1.50247.0001 1.50248.0001 1.50249.0001 1.50267.0001 1.50268.0001 1.50269.0001 1.50356.0001 1.50357.0001
HPLC columns, Purospher® STAR Si and NH₂
Kromatografikolonner
Purospher® STAR HPLC columns based on highest purity silica are also available for normal phase separations.
Purospher® STAR Si (silica) offers the highest separation efficiency for normal phase chromatography of low molecular weight compounds soluble in organic solvents.
Purospher® STAR NH₂ (amino phase) is primarily designed for carbohydrate analysis with a typical mobile phase consisting of acetonitrile and water. Additionally, Purospher® STAR NH₂ can also be used in the normal phase retention mode.
These LiChroCART® cartridges require manu-CART® cartridge columnMore Product Information