Eddikesyre 75% ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

1.37011.1000 1.37011.9025 1.37011.9950
1.37011.1000EA 1190 DKK
1.37011.1000 1.37011.9025 1.37011.9950
Eddikesyre 75% ≥75%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Eddikesyre

Advarsel: Store at +15°C to +25°C.

Formel: H₃CCOOH
Molekylvægt: 60.05 g/mol
Kogepunkt: 118
Smeltepunkt: 17
Flammepunkt: 38,5
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
UN nummer: 2790
ADR: 8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR