Citronsyre monohydrat 99.5-100.5% (alkalimetric, calculated on anhydrous substance), crystals, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

1.37003.5000 1.37003.9012 1.37003.9029
1.37003.5000EA 5860 DKK
1.37003.5000 1.37003.9012 1.37003.9029
Citronsyre monohydrat 99.5-100.5% (alkalimetric, calculated on anhydrous substance), crystals, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®
Citronsyre monohydrat

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certificeringer: Corresponds to Ph. Eur., BP, JP, USP, ACS

Formel: C₆H₈O₇.H₂O
Smeltepunkt: 135…152 °C
Massefylde: 1,552 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149972
CAS-nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (alkalimetric, calculated on anhydrous substance) 99.5 - 100.5 %
Identity (IR-spectrum) passes test
Appearance white to almost white or colorless crystals
Appearance of solution (200 g/l; water) clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
In water insoluble matter # ≤ 0.005 %
Chlorid (Cl) # ≤ 0.001 %
Phosphat (PO4) # ≤ 0.001 %
Sulfate (SO₄) ≤ 20 ppm
Heavy metals (as Pb) ≤ 10 ppm
Al (Aluminium) ≤ 0.2 ppm
Fe (Iron) # ≤ 3 ppm
Pb (Lead) # ≤ 2 ppm
Oxalates (as C₂H₂O₄) ≤ 360 ppm
Residual solvents (ICH (Q3C)) excluded by manufacturing process
Readily carbonisable substance passes test
Sulfated ash (600 °C) ≤ 0.02 %
Water (according to Karl Fischer) 7.5 - 9.0 %
Total aerobic microbial count (TAMC) ≤ 100 CFU/g
Total combined yeasts/moulds count (TYMC) ≤ 10 CFU/g
Bile-tolerant gram-negative bacteria (absent in 1 g) passes test
Candida albicans (absent in 1 g) passes test
Escherichia coli (absent in 1 g) passes test
Pseudomonas aeruginosa (absent in 1 g) passes test
Salmonella (absent in 10 g) passes test
Staphylococcus aureus (absent in 1 g) passes test
Bacterial endotoxins < 10
# Additional requirements by ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR