Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.16755.1000
1.16755.1000EA 899.9 DKK
1.16755.1000
Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Buffere Ioniseringsbuffere
10 g/L CsCl + 100 g/L La in dilute hydrochloric acid.