Reflectoquant® teststrips, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.16127.0001
1.16127.0001EA 1250 DKK
1.16127.0001
Reflectoquant® teststrips, Supelco®
Miljø test kit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
The test measures total lactic acid (sum of D- and L-lactic acid in beverages, e.g. wine, beer and fruit/vegetable juice after dilution. Furthermore the test can be used for the determination of lactic acid in food after pretreatment (typically dilution).