Reagent Kits til Protein og Fibre Analyser
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.12979.0001
1.12979.0001EA 5950 DKK
1.12979.0001
Reagent Kits til Protein og Fibre Analyser
Fødevaretest systemer
Simple reagent kits, used in food analysis, that use colour chemistry to determine levels of protein and fibre. Please note these kits require the use of a photometer or spectrophotometer.