N-Acetyl-DL-tryptophan, EMPROVE® EXPERT BP, Ph. Eur., extra pure for biopharmaceutical production low in endotoxins, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ac-DL-Trp-OH

1.12488.1000 1.12488.9025
1.12488.1000EA 25080 DKK
1.12488.1000 1.12488.9025
N-Acetyl-DL-tryptophan, EMPROVE® EXPERT BP, Ph. Eur., extra pure for biopharmaceutical production low in endotoxins, SAFC®
N-Acetyl-DL-tryptophan

N-Acetyl-DL-tryptophan is suitable for biopharmaceutical production.

Formel: C₁₃H₁₄N₂O₃
Smeltepunkt: 205…206 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005644
CAS-nummer: 87-32-1
EINECS: 201-739-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR