Calciumcarbonat, EMPROVE® ESSENTIAL FCC, E170, BP, Ph. Eur., JP, USP suitable for use as excipient Al_0,0001, precipitated, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Marble

1.12120.9029
1.12120.9029EA 0 DKK
1.12120.9029
Calciumcarbonat, EMPROVE® ESSENTIAL FCC, E170, BP, Ph. Eur., JP, USP suitable for use as excipient Al_0,0001, precipitated, SAFC®
Calcium carbonate
Formel: CaCO₃
Molekylvægt: 100.09 g/mol
Smeltepunkt: 825
Massefylde: 2.71 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010906
CAS-nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR