Chrom, powder, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


M-Clarity™ quality level = MQ200

1.12097.0500 1.12097.5000
1.12097.0500EA 1520 DKK
1.12097.0500 1.12097.5000
Chrom, powder, Sigma-Aldrich®
Chromium
Formel: Cr
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010944
CAS-nummer: 7440-47-3
EINECS: 231-157-5
UN nummer: 3089
ADR: 4.1,III

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) insoluble
Melting Point 1890 °C
Molar Mass 51.99 g/mol
Boiling Point 2670 °C (1013 hPa)
Density 7.20 g/cm3 (20 °C)
pH (H2O) not applicable

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR