Natriumchlorid 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Saline, NaCl, Sea salt, Halite

M-Clarity™ quality level = MQ100

1.09945.0001
1.09945.0001EA 240 DKK
1.09945.0001
Natriumchlorid 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Sodium chloride
Formel: NaCl
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Density 1.08 g/cm3 (20 °C)
pH 4.8 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR