Kaliumhydroxid 1 mol/l aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potash caustic, Caustic potash

MQ100

1.09918.0001
1.09918.0001EA 322 DKK
1.09918.0001
Kaliumhydroxid 1 mol/l aqueous solution, Titrisol®, Supelco®
Potassium hydroxide
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 1320
Smeltepunkt: 360
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 1814
ADR: 8
Merck Index: 12,07806

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Density 1.58 g/cm3 (20 °C)
pH 13.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR