Buffers, concentrated solutions, Titrisol®, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

1.09879.0001 1.09880.0001 1.09881.0001 1.09882.0001 1.09883.0001 1.09884.0001 1.09885.0001 1.09886.0001 1.09887.0001 1.09888.0001 1.09889.0001 1.09890.0001 1.09892.0001 1.09893.0001 1.09883.0050 1.09884.0050 1.09887.0050 1.09889.0050
1.09879.0001EA 300 DKK
1.09879.0001 1.09880.0001 1.09881.0001 1.09882.0001 1.09883.0001 1.09884.0001 1.09885.0001 1.09886.0001 1.09887.0001 1.09888.0001 1.09889.0001 1.09890.0001 1.09892.0001 1.09893.0001 1.09883.0050 1.09884.0050 1.09887.0050 1.09889.0050
Buffers, concentrated solutions, Titrisol®, Supelco®
Buffere pH buffere

Titrisol® buffer concentrate for 500 ml buffer solution [20 °C].


  • Traceable to secondary reference material (SRM) of NIST and PTB
  • Buffer concentrate for the preparation of 500 ml of buffer solution
  • Accuracy: ≤0,05 pH units at 20 °C


* according to Wiese for 1000 ml solution

Bestil nu

Specifikationer

M-Clarity quality level MQ100
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR