2-Propanol, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopropylalkohol

MQ300

1.09634.1000 1.09634.1011 1.09634.2500 1.09634.2511 1.09634.5000 1.09634.6010 1.09634.6025 1.09634.6190 1.09634.9180 1.09634.9950 1.09634.4000
1.09634.1000EA 201.15 DKK
1.09634.1000 1.09634.1011 1.09634.2500 1.09634.2511 1.09634.5000 1.09634.6010 1.09634.6025 1.09634.6190 1.09634.9180 1.09634.9950 1.09634.4000
2-Propanol, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®
2-Propanol

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: (CH₃)₂CHOH
Molekylvægt: 60.1 g/mol
Kogepunkt: 81… 83 °C (1013 hPa)
Smeltepunkt: –89 °C
Massefylde: 0.784 …0.787 g/cm³ (20 °C)
Flammepunkt: 12 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011674
CAS-nummer: 67-63-0
EINECS: 200-661-7
UN nummer: 1219
ADR: 3
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

På tilbud lige nu

De valgte varer nedenfor er i øjeblikket på udsalg

Bestil nu

Specifikationer Testresultater

Purity (GC) ≥ 99.8 %
Identity (IR) conforms
Appearance clear
Color ≤ 10 Hazen
Solubility in water conforms
Acidity ≤ 0.0001 meq/g
Alkalinity ≤ 0.0001 meq/g
Density (d 20 °C/20 °C) 0.784 - 0.787
Boiling point 81 - 83 °C
Chloride (Cl) ≤ 300 ppb
Nitrate (NO₃) ≤ 300 ppb
Phosphate (PO₄) ≤ 500 ppb
Sulfate (SO₄) ≤ 1000 ppb
Substances reducing potassium permanganate (as O) ≤ 2 ppm
Carbonyl compounds (as CO) ≤ 0.002 %
Matter discolored by H₂SO₄ ≤ 10 Hazen
Acetone (GC) ≤ 0.01 %
Ethanol (GC) ≤ 0.01 %
Isopropylether (GC) ≤ 0.01 %
Methanol (GC) ≤ 0.01 %
1-Propylalcohol (GC) ≤ 0.1 %
Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.000002 %
Au (Gold) ≤ 0.000002 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Be (Beryllium) ≤ 0.000002 %
Bi (Bismuth) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Ga (Gallium) ≤ 0.000002 %
In (Indium) ≤ 0.000002 %
Li (Lithium) ≤ 0.000005 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Mo (Molybdenum) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Pt (Platinum) ≤ 0.000002 %
Sb (Antimony) ≤ 0.000002 %
Sn (Tin) ≤ 0.00001 %
Ti (Titanium) ≤ 0.000002 %
Tl (Thallium) ≤ 0.000002 %
V (Vanadium) ≤ 0.000002 %
Zn (Zinc) ≤ 0.00001 %
Zr (Zirconium) ≤ 0.000002 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.05 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR