Zinkoxid Reag. Ph. Eur., ACS analytical reagent, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Zinci oxidum, Zinc (II) oxide

M-Clarity™ quality level = MQ300

1.08849.0500 1.08849.1000 1.08849.9025
1.08849.0500EA 596 DKK
1.08849.0500 1.08849.1000 1.08849.9025
Zinkoxid Reag. Ph. Eur., ACS analytical reagent, Supelco®
Zinkoxid
Formel: ZnO
Smeltepunkt: 1975 °C
Massefylde: 5,61 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011300
CAS-nummer: 1314-13-2
EINECS: 215-222-5
UN nummer: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) insoluble
Melting Point 1975 °C
Molar Mass 81.37 g/mol
Bulk Density 200 - 700 kg/m3
Density 5.61 g/cm3 (20 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR