EDTA trikaliumsalt dihydrat, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate

M-Clarity™ quality level = MQ100

1.08445.0250
1.08445.0250EA 2350 DKK
1.08445.0250
EDTA trikaliumsalt dihydrat, Supelco®
EDTA trikaliumsalt dihydrat
Formel: C₁₀H₁₃K₃N₂O₈·2H₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149672
CAS-nummer: 65501-24-8
EINECS: 241-543-5

Bestil nu

Specifikationer Testresultater

Melting Point 180 - 185 °C
Molar Mass 442.56 g/mol
pH 8 - 9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR