Thiamin hydrochlorid, Millipore®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

M-Clarity™ quality level = MQ100

1.08181.0025 1.08181.0250
1.08181.0025EA 383 DKK
1.08181.0025 1.08181.0250
Thiamin hydrochlorid, Millipore®
Thiamin hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Smeltepunkt: 252 °C
Massefylde: 1,401 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012780
CAS-nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR