Tetrahydrofuran, EMPLURA®, extra pure, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Butylene oxide, THF, Diethylene oxide

M-Clarity™ quality level = MQ200

1.08114.1000 1.08114.2500 1.08114.6025 1.08114.6190 1.08114.9190
1.08114.1000EA 595 DKK
1.08114.1000 1.08114.2500 1.08114.6025 1.08114.6190 1.08114.9190
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Tetrahydrofuran

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formel: C₄H₈O
Kogepunkt: 66 °C (1013 hPa)
Massefylde: 0,888 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005356
CAS-nummer: 109-99-9
EINECS: 203-726-8
UN nummer: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR