Strontiumchlorid hexahydrat, EMSURE® ACS for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ300

1.07865.0250 1.07865.1000 1.07865.9050
1.07865.0250EA 711 DKK
1.07865.0250 1.07865.1000 1.07865.9050
Strontiumchlorid hexahydrat, EMSURE® ACS for analysis, Supelco®
Strontiumchlorid hexahydrat
Formel: SrCl₂·6H₂O
Kogepunkt: 100 °C (1013 hPa)
Smeltepunkt: 61 °C
Massefylde: 1,93 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149865
CAS-nummer: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Bestil nu

Specifikationer Testresultater

Solubility 2058 g/l (40 °C)
Melting Point 61 °C
Molar Mass 266.62 g/mol
Bulk Density 1050 kg/m3
Boiling Point 100 °C
Density 1.95 g/cm3 (20 °C)
pH 5 - 7 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR