Hav sand, extra pure, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


M-Clarity™ quality level = MQ200

1.07711.1000 1.07711.5000 1.07711.9025
1.07711.1000EA 883 DKK
1.07711.1000 1.07711.5000 1.07711.9025
Hav sand, extra pure, Supelco®
Hav sand
Formel: SiO₂
Kogepunkt: 2230 °C (1013 hPa)
Smeltepunkt: 1710 °C
Massefylde: 2,66 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 14808-60-7
EINECS: 231-545-4

Bestil nu

Specifikationer Testresultater

Substances soluble in hydrochloric acid ≤ 0.5 %
Chloride (Cl) ≤ 0.01 %
Loss on ignition (900 °C) ≤ 0.5 %
Particle size (0.1 - 0.315 mm) ≥ 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR