D(-)-Ribose for biochemistry, Millipore®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


M-Clarity™ quality level = MQ100

1.07605.0050
1.07605.0050EA 1090 DKK
1.07605.0050
D(-)-Ribose for biochemistry, Millipore®
D(-)-Ribose
Formel: C₅H₁₀O₅
Kogepunkt: 331 °C (1013 hPa)
Smeltepunkt: 90…95 °C
Massefylde: 0,8 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00135453
CAS-nummer: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR